• USA Home
  • 07973 - 2-Amino-5-fluorobenzoic acid

EMAIL THIS PAGE TO A FRIEND
07973 Sigma-Aldrich

2-Amino-5-fluorobenzoic acid

purum, ≥97.0% (T)

Synonym: 5-Fluoroanthranilic acid

  • CAS Number 446-08-2

  • Linear Formula H2NC6H3(F)CO2H

  • Molecular Weight 155.13

  •  Beilstein Registry Number 2803664

  •  EC Number 207-159-7

  •  MDL number MFCD00055566

  •  PubChem Substance ID 24846134

Purchase

Properties

Related Categories Aromatic Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   FPQMGQZTBWIHDN-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (T)
mp   181-183 °C(lit.)
  183-188 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?