• USA Home
  • 08035 - L-2-Amino-3-guanidinopropionic acid hydrochloride

EMAIL THIS PAGE TO A FRIEND
08035 Sigma-Aldrich

L-2-Amino-3-guanidinopropionic acid hydrochloride

purum, ≥98.0% (CHN)

Synonym: L-α-Amino-β-guanidinopropionic acid hydrochloride

  • CAS Number 1482-99-1

  • Empirical Formula (Hill Notation) C4H10N4O2 · HCl

  • Molecular Weight 182.61

  •  Beilstein Registry Number 6317820

  •  EC Number 216-044-0

  •  MDL number MFCD00036989

Purchase

Properties

Related Categories Arginine Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   ZOXYHKHLLCDEAX-DKWTVANSSA-N
grade   purum
assay   ≥98.0% (CHN)
mp   ~220 °C (dec.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?