• USA Home
  • 08045 - 7-Aminoheptanoic acid

EMAIL THIS PAGE TO A FRIEND
08045 Sigma-Aldrich

7-Aminoheptanoic acid

purum, ≥97.0% (CHN)

Synonym: 7-Aminooenanthic acid

  • CAS Number 929-17-9

  • Linear Formula H2N(CH2)6CO2H

  • Molecular Weight 145.20

  •  Beilstein Registry Number 906887

  •  EC Number 213-197-5

  •  MDL number MFCD00008242

  •  PubChem Substance ID 329748542

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Linear Core Amino Acids, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   XDOLZJYETYVRKV-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (CHN)
mp   192-195 °C(lit.)
  ~195 °C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
MJ1770000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

284637 7-Aminoheptanoic acid, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?