• USA Home
  • 08280 - α-Aminoisobutyric acid

EMAIL THIS PAGE TO A FRIEND
08280 Sigma-Aldrich

α-Aminoisobutyric acid

puriss., ≥99.0% (NT)

Synonym: 2-Methylalanine, Aib, alpha-Aminoisobutyric acid

  • CAS Number 62-57-7

  • Linear Formula (CH3)2C(NH2)COOH

  • Molecular Weight 103.12

  •  Beilstein Registry Number 506496

  •  EC Number 200-544-0

  •  MDL number MFCD00008049

  •  PubChem Substance ID 329748563

Purchase

Properties

Related Categories A - H, Alanine Derivatives, Amino Acids, Biochemicals and Reagents, Chemical Biology,
InChI Key   FUOOLUPWFVMBKG-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (NT)
ign. residue   ≤0.3%
mp   ≥300 °C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
AY7000000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

850993 2-Aminoisobutyric acid, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?