• USA Home
  • 08290 - DL-β-Aminoisobutyric acid

EMAIL THIS PAGE TO A FRIEND
08290 Sigma-Aldrich

DL-β-Aminoisobutyric acid

puriss., ≥99.0% (NT)

Synonym: α-Methyl-β-alanine

  • CAS Number 144-90-1

  • Linear Formula NH2CH2-CH(CH3)COOH

  • Molecular Weight 103.12

  •  Beilstein Registry Number 1720958

  •  EC Number 205-644-8

  •  MDL number MFCD00008145

  •  PubChem Substance ID 329748564

Purchase

Properties

Related Categories Beta-Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   QCHPKSFMDHPSNR-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (NT)
ign. residue   ~0.1%
mp   178-180 °C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

217794 DL-3-Aminoisobutyric acid, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?