• USA Home
  • 08329 - 4-Amino-3-methylphenol

EMAIL THIS PAGE TO A FRIEND
08329 Sigma-Aldrich

4-Amino-3-methylphenol

purum, ≥98.0% (NT)

Synonym: 2-Amino-5-hydroxytoluene, 4-Hydroxy-2-methylaniline, 4-Amino-m-cresol

  • CAS Number 2835-99-6

  • Linear Formula H2NC6H3(CH3)OH

  • Molecular Weight 123.15

  •  Beilstein Registry Number 2078803

  •  EC Number 220-621-2

  •  MDL number MFCD00007871

  •  PubChem Substance ID 24846159

Purchase

Properties

Related Categories Building Blocks, C6 to C8, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds,
InChI Key   QGNGOGOOPUYKMC-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (NT)
mp   176-179 °C(lit.)
  177-179 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
GO6826000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?