• USA Home
  • 08332 - 2-Amino-4-methylphenol

EMAIL THIS PAGE TO A FRIEND
08332 Sigma-Aldrich

2-Amino-4-methylphenol

technical, ≥98.0% (NT)

Synonym: 2-Hydroxy-5-methylaniline, 2-Amino-p-cresol, 3-Amino-4-hydroxytoluene

  • CAS Number 95-84-1

  • Linear Formula H2NC6H3(CH3)OH

  • Molecular Weight 123.15

  •  Beilstein Registry Number 606494

  •  EC Number 202-457-3

  •  MDL number MFCD00007699

  •  PubChem Substance ID 329748568

Purchase

Properties

InChI Key   ZMXYNJXDULEQCK-UHFFFAOYSA-N
grade   technical
assay   ≥98.0% (NT)
mp   133-136 °C(lit.)
  135-138 °C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
2
RTECS 
SJ6078000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?