• USA Home
  • 08336 - 2-Amino-5-methylphenol

EMAIL THIS PAGE TO A FRIEND
08336 Sigma-Aldrich

2-Amino-5-methylphenol

purum, ≥97.0% (NT/TLC)

Synonym: 2-Hydroxy-4-methylaniline, 4-Amino-3-hydroxytoluene, 6-Amino-m-cresol

Purchase

Properties

InChI Key   HCPJEHJGFKWRFM-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (NT/TLC)
mp   159-162 °C(lit.)
  162-166 °C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

144916 2-Amino-5-methylphenol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?