• USA Home
  • 08399 - (S)-(+)-1-Methoxy-3-phenyl-2-propylamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
08399 Sigma-Aldrich

(S)-(+)-1-Methoxy-3-phenyl-2-propylamine hydrochloride

purum, ≥98.0% (AT)

Synonym: (+)-O-Methyl-L-phenylalaninol hydrochloride, (S)-(+)-2-Amino-1-methoxy-3-phenylpropane hydrochloride

  • CAS Number 64715-81-7

  • Empirical Formula (Hill Notation) C10H15NO · HCl

  • Molecular Weight 201.69

  •  Beilstein Registry Number 5139125

  •  MDL number MFCD00043238

  •  PubChem Substance ID 24846169

Purchase

Properties

Related Categories Amine Salts, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   FTMKEFCCBFJBJI-PPHPATTJSA-N
grade   purum
assay   ≥98.0% (AT)
optical activity   [α]20/D +20±2°, c = 2.5% in ethanol
mp   149-151 °C

Description

Other Notes

Chiral amine as auxiliary reagent for an asymmetric synthesis of α-alkyl and α,α-dialkyl cyclic ketones

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?