EMAIL THIS PAGE TO A FRIEND
08436 Sigma-Aldrich

Butyronitrile

analytical standard

Synonym: Propyl cyanide

  • CAS Number 109-74-0

  • Linear Formula CH3CH2CH2CN

  • Molecular Weight 69.11

  •  Beilstein Registry Number 1361452

  •  EC Number 203-700-6

  •  MDL number MFCD00001968

  •  PubChem Substance ID 24846181

Purchase

Properties

Related Categories A-B, A-C, Additional Standards, Alpha Sort, Analytical Standards,
grade   analytical standard
vapor density   2.4 (vs air)
vapor pressure   23 mmHg ( 25 °C)
InChI Key   KVNRLNFWIYMESJ-UHFFFAOYSA-N
assay   ≥99.5% (GC)
autoignition temp.   910 °F
shelf life   limited shelf life, expiry date on the label
refractive index   n20/D 1.384(lit.)
  n20/D 1.384
bp   115-117 °C(lit.)
mp   −112 °C(lit.)
density   0.794 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2411 6.1(3) / PGII
WGK Germany 
1
RTECS 
ET8750000
Flash Point(F) 
64.4 °F
Flash Point(C) 
18 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?