• USA Home
  • 08460 - Cyclopropanemethylamine

EMAIL THIS PAGE TO A FRIEND
08460 Sigma-Aldrich

Cyclopropanemethylamine

purum, ≥96.0% (GC)

Synonym: (Aminomethyl)cyclopropane

  • CAS Number 2516-47-4

  • Empirical Formula (Hill Notation) C4H9N

  • Molecular Weight 71.12

  •  Beilstein Registry Number 773674

  •  EC Number 219-737-6

  •  MDL number MFCD00037147

  •  PubChem Substance ID 329748588

Purchase

Properties

InChI Key   IGSKHXTUVXSOMB-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (GC)
refractive index   n20/D 1.433(lit.)
bp   83-86 °C(lit.)
density   0.83 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2733PSN1 8(3) / PGII
WGK Germany 
3
Flash Point(F) 
35.6 °F
Flash Point(C) 
2 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?