• USA Home
  • 08545 - Aniline phthalate solution

EMAIL THIS PAGE TO A FRIEND
08545 Sigma-Aldrich

Aniline phthalate solution

~0.1 M in butanol/water, for TLC derivatization

Synonym: Phthalic acid Aniline salt

  • CAS Number 50930-79-5

  • Empirical Formula (Hill Notation) C14H13NO4

  • Molecular Weight 259.26

  •  Beilstein Registry Number 3777765

  •  MDL number MFCD00042034

  •  PubChem Substance ID 329748599

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Derivatization Reagents, Derivatization Reagents TLC, Sugars,
grade   for TLC derivatization
InChI Key   QGPZXNSBZMHHSR-UHFFFAOYSA-N
concentration   ~0.1 M in butanol/water
density   0.857 g/mL at 20 °C
suitability   corresponds for TLC
storage temp.   2-8°C

Description

General description

Visit our Titration Center to learn more.

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN1120 - class 3 - PG 3 - Butanols solution
WGK Germany 
2
Flash Point(F) 
104 °F
Flash Point(C) 
40 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?