• USA Home
  • 08557 - (Aminomethyl)polystyrene

EMAIL THIS PAGE TO A FRIEND
08557 Sigma-Aldrich

(Aminomethyl)polystyrene

100-200 mesh, extent of labeling: ~2 mmol/g amine loading

Synonym: AM-polystyrene, Benzylamine polymer-bound, Poly(styrene-co-divinylbenzene), aminomethylated, Poly(styrene-co-vinylbenzylamine-co-divinylbenzene, Polystyrene crosslinked with divinylbenzene, aminomethylated

Purchase

Properties

Related Categories Base Resins for SPPS, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Polymer Supports for SPPS More...
extent of labeling   ~2 mmol/g amine loading
matrix   crosslinked with 3% DVB
particle size   100-200 mesh
storage temp.   2-8°C

Description

Application

Scavenges acids, acid chlorides, anhydrides, aldehydes and other electrophiles

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?