• USA Home
  • 08580 - 2-Amino-2-methyl-1-propanol

EMAIL THIS PAGE TO A FRIEND
08580 Sigma-Aldrich

2-Amino-2-methyl-1-propanol

purum, ≥97.0% (GC)

Synonym: β-Aminoisobutyl alcohol, AMP 95, AMP

  • CAS Number 124-68-5

  • Linear Formula (CH3)2C(NH2)CH2OH

  • Molecular Weight 89.14

  •  Beilstein Registry Number 505979

  •  EC Number 204-709-8

  •  MDL number MFCD00008051

  •  PubChem Substance ID 329748612

Purchase

Properties

Related Categories Amino Alcohols, Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds More...
vapor density   3 (vs air)
vapor pressure   <1 mmHg ( 25 °C)
InChI Key   CBTVGIZVANVGBH-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.4455(lit.)
useful pH range   9.0 - 10.5
pKa (25 °C)   9.7
bp   165 °C(lit.)
mp   24-28 °C(lit.)
density   0.934 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Flash Point(F) 
154.4 °F
Flash Point(C) 
68 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?