• USA Home
  • 08585 - 2-Amino-2-methyl-1-propanol hydrochloride

EMAIL THIS PAGE TO A FRIEND
08585 Sigma-Aldrich

2-Amino-2-methyl-1-propanol hydrochloride

≥99.0% (AT)

Synonym: β-Aminoisobutyl alcohol hydrochloride, AMP

  • CAS Number 3207-12-3

  • Linear Formula NH2C(CH3)2CH2OH · HCl

  • Molecular Weight 125.60

  •  Beilstein Registry Number 3668007

  •  EC Number 221-713-5

  •  MDL number MFCD00039086

  •  PubChem Substance ID 329748615

Purchase

Properties

InChI Key   PMZIUAOBHNJYQT-UHFFFAOYSA-N
assay   ≥99.0% (AT)
impurities   ≤0.2% water
useful pH range   9.0 - 10.5
pKa (25 °C)   9.7
mp   204-206 °C
solubility   H2O: soluble0.5 g/mL, clear, colorless

Description

Other Notes

Biological buffer component

Application

2-Amino-2-methyl-1-propanol hydrochloride has been used as a wash buffer in ELISA. It has also been used for preparing buffer for fiber typing by myofibrillar actomyosin ATPase histochemistry.

Biochem/physiol Actions

2-Amino-2-methyl-1-propanol is an alkalizing agent that maintains the pH of a system.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A5888 2-Amino-2-methyl-1-propanol hydrochloride

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?