• USA Home
  • 08604 - Cellulose acetate sheets

EMAIL THIS PAGE TO A FRIEND
08604 Sigma-Aldrich

Cellulose acetate sheets

perforated, size 25 mm × 150 mm

Purchase

Properties

Related Categories Cellulose Electrophoresis, Molecular Biology, Protein Electrophoresis, Proteomics
quality   perforated
size   25 mm × 150 mm

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Features and Benefits

With cellulose acetate electrophoresis, you can separate nearly all sample materials that migrate in an electrical field. Apply 0.06 to 0.15 mg of sample mixture per cm application line. Concentrate the mixture to at least 1 mg/ml. Advantages of those strips include:
• High porosity of the strip (approx. 80 % free pore volume)
• Isotropic, integral matrix for ideal flow conditions
• Minimal diffusion effect and low adsorption capacity
• Perfect transparency because of the pore system
• Ideal for the non-toxic ATX reagent system
• Virtually unlimited stability at room temperature
• Two identical sides, no need to search for the "right" side
Available in different sizes for all current chamber types

These strips can also be used for high-voltage electrophoresis, chromatography, and immunodiffusion.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?