• USA Home
  • 08660 - 3-Amino-2-naphthoic acid

EMAIL THIS PAGE TO A FRIEND
08660 Sigma-Aldrich

3-Amino-2-naphthoic acid

technical, ≥80% (NT)

Purchase

Properties

Related Categories Aromatic Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   XFXOLBNQYFRSLQ-UHFFFAOYSA-N
grade   technical
assay   ≥80% (NT)
mp   212-215 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
QL1400000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?