EMAIL THIS PAGE TO A FRIEND
08970 Sigma-Aldrich

Butyl vinyl ether

analytical standard

Synonym: (Butyloxy)ethylene, Vinyl butyl ether

  • CAS Number 111-34-2

  • Linear Formula CH3(CH2)3OCH=CH2

  • Molecular Weight 100.16

  •  Beilstein Registry Number 1560217

  •  EC Number 203-860-7

  •  MDL number MFCD00009454

  •  PubChem Substance ID 329748650

Purchase

Properties

Related Categories A-B, A-C, Additional Standards, Alpha Sort, Analytical Standards,
grade   analytical standard
vapor density   3.5 (vs air)
vapor pressure   42 mmHg ( 20 °C)
InChI Key   UZKWTJUDCOPSNM-UHFFFAOYSA-N
assay   ≥99.5% (GC)
autoignition temp.   437 °F
shelf life   limited shelf life, expiry date on the label
contains   ~0.1% potassium hydroxide as stabilizer
refractive index   n20/D 1.400(lit.)
  n20/D 1.401
bp   94 °C(lit.)
mp   −92 °C(lit.)
density   0.774 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
May form explosive peroxides.
RIDADR 
UN 2352 3 / PGII
WGK Germany 
2
RTECS 
KN5950000
Flash Point(F) 
10.4 °F
Flash Point(C) 
-12 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?