EMAIL THIS PAGE TO A FRIEND
09120 Sigma-Aldrich

2-Aminophenol

purum, ≥98.0% (HPLC)

  • CAS Number 95-55-6

  • Linear Formula H2NC6H4OH

  • Molecular Weight 109.13

  •  Beilstein Registry Number 606075

  •  EC Number 202-431-1

  •  MDL number MFCD00007690

  •  PubChem Substance ID 329748692

Purchase

Properties

Related Categories Building Blocks, C6 to C8, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds,
InChI Key   CDAWCLOXVUBKRW-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   170-175 °C(lit.)
  170-175 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2512 6.1 / PGIII
WGK Germany 
2
RTECS 
SJ4950000
Flash Point(F) 
334.4 °F
Flash Point(C) 
168 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A71301 2-Aminophenol, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?