EMAIL THIS PAGE TO A FRIEND
09130 Sigma-Aldrich

3-Aminophenol

purum, ≥98.0% (T)

  • CAS Number 591-27-5

  • Linear Formula H2NC6H4OH

  • Molecular Weight 109.13

  •  Beilstein Registry Number 636059

  •  EC Number 209-711-2

  •  MDL number MFCD00007786

  •  PubChem Substance ID 24846252

Purchase

Properties

Related Categories Building Blocks, C6 to C8, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds,
InChI Key   CWLKGDAVCFYWJK-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
  ≥98.0% (T)
bp   164 °C/11 mmHg(lit.)
mp   120-124 °C(lit.)
  120-124 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2512 6.1 / PGIII
WGK Germany 
2
RTECS 
SJ4900000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?