• USA Home
  • 09187 - ATX Clear, ready-to-use solution

EMAIL THIS PAGE TO A FRIEND
09187 Sigma-Aldrich

ATX Clear, ready-to-use solution

Synonym: Phero Clear

Purchase

Description

Packaging

6 bottles with 1 L ready-to-use solution (each)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
1
Flash Point(F) 
141.8 °F
Flash Point(C) 
61 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?