• USA Home
  • 09230 - 4-(6-Methyl-2-benzothiazolyl)aniline

EMAIL THIS PAGE TO A FRIEND
09230 Sigma-Aldrich

4-(6-Methyl-2-benzothiazolyl)aniline

technical, ≥98.0% (NT)

Synonym: 2-(4-Aminophenyl)-6-methylbenzothiazole, Dehydrothiotoluidine

  • CAS Number 92-36-4

  • Empirical Formula (Hill Notation) C14H12N2S

  • Molecular Weight 240.32

  •  Beilstein Registry Number 175967

  •  EC Number 202-150-4

  •  MDL number MFCD00005780

  •  PubChem Substance ID 329748743

Purchase

Properties

Related Categories Building Blocks, C12 to C20+, Chemical Synthesis, Heterocyclic Building Blocks, Thiazoles More...
InChI Key   XRTJYEIMLZALBD-UHFFFAOYSA-N
grade   technical
assay   ≥98.0% (NT)
bp   434 °C(lit.)
mp   188-193 °C(lit.)
  188-193 °C

Description

Caution

may discolor to brown on storage

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
DL2820000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?