EMAIL THIS PAGE TO A FRIEND
09253 Sigma-Aldrich

Luminol

for chemiluminescence, ≥98.0% (HPLC)

Synonym: 3-Aminophthalhydrazide, 5-Amino-2,3-dihydro-1,4-phthalazinedione

  • CAS Number 521-31-3

  • Empirical Formula (Hill Notation) C8H7N3O2

  • Molecular Weight 177.16

  •  Beilstein Registry Number 383929

  •  EC Number 208-309-4

  •  MDL number MFCD00006890

  •  PubChem Substance ID 329748748

Purchase

Properties

grade   for chemiluminescence
InChI Key   HWYHZTIRURJOHG-UHFFFAOYSA-N
assay   ≥98.0% (HPLC)
mp   >300 °C(lit.)

Description

Analysis Note

luminescence test: Emmax 440 nm (chemiluminescence; in 60 mM K2S2O8, 100 mM K2CO3, pH 11.5; after addition of H2O2)

Features and Benefits

Used for chemiluminescence analysis of, for example, metal cations, blood, and glucococorticoids.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
TH8890060

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?