• USA Home
  • 09281 - (R)-(−)-1-Amino-2-propanol

EMAIL THIS PAGE TO A FRIEND
09281 Sigma-Aldrich

(R)-(−)-1-Amino-2-propanol

purum, ≥98.0% (sum of enantiomers, GC)

Synonym: (−)-Isopropanolamine

  • CAS Number 2799-16-8

  • Linear Formula CH3CH(OH)CH2NH2

  • Molecular Weight 75.11

  •  Beilstein Registry Number 1718869

  •  EC Number 220-532-9

  •  MDL number MFCD00064428

  •  PubChem Substance ID 24846300

Purchase

Properties

Related Categories Amino Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Chiral Resolution Reagents,
vapor density   2.6 (vs air)
vapor pressure   <1 mmHg ( 20 °C)
InChI Key   HXKKHQJGJAFBHI-GSVOUGTGSA-N
grade   purum
assay   ≥98.0% (sum of enantiomers, GC)
optical activity   [α]/D −23.5±2°, c = 1% in methanol
autoignition temp.   635 °F
impurities   ≤1% water
refractive index   n20/D 1.448
  n20/D 1.4482(lit.)
bp   160 °C(lit.)
mp   24-26 °C(lit.)
density   0.954 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Used for the preparation of chiral enamines; Synthesis of 2-methylaziridine; Use as chiral ligand

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3259PSN1 8 / PGII
WGK Germany 
1
Flash Point(F) 
159.8 °F
Flash Point(C) 
71 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?