• USA Home
  • 09297 - Aminopropyl silica gel

EMAIL THIS PAGE TO A FRIEND
09297 Sigma-Aldrich

Aminopropyl silica gel

for preparative liquid chromatography, normal phase, weak anion exchanger

Synonym: Aminopropyl silanised silica gel, Silica gel NH2

Purchase

Properties

Related Categories Aminopropyl, Analytical/Chromatography, Anion, LPLC Separation Media, LPLC/TLC/Paper Chromatography More...
grade   for preparative liquid chromatography
particle size   15-35 μm
pore size   ~9 nm pore size

Description

General description

reversed phase of polar compounds as carbohydrates

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?