• USA Home
  • 09298 - Aminopropyl silica gel

EMAIL THIS PAGE TO A FRIEND
09298 Sigma-Aldrich

Aminopropyl silica gel

for preparative liquid chromatography, extent of labeling: ~0.9 mmol/g amino groups loading

Synonym: Aminopropyl silanised silica gel, Silica gel NH2

Purchase

Properties

Related Categories Aminopropyl, Analytical/Chromatography, Anion, LPLC Separation Media, LPLC/TLC/Paper Chromatography More...
grade   for preparative liquid chromatography
extent of labeling   ~0.9 mmol/g amino groups loading
particle size   40-63 μm
pore size   ~9 nm pore size

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?