EMAIL THIS PAGE TO A FRIEND
09332 Sigma-Aldrich

Aminopyrazine

purum, ≥97.0% (NT)

Synonym: Pyrazinamine

  • CAS Number 5049-61-6

  • Empirical Formula (Hill Notation) C4H5N3

  • Molecular Weight 95.10

  •  Beilstein Registry Number 107025

  •  EC Number 225-748-7

  •  MDL number MFCD00006137

  •  PubChem Substance ID 24846315

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Pyrazines
InChI Key   XFTQRUTUGRCSGO-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (NT)
mp   118-120 °C(lit.)
  118-120 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?