• USA Home
  • 09371 - 1-Bromo-4-isopropylbenzene

EMAIL THIS PAGE TO A FRIEND
09371 Sigma-Aldrich

1-Bromo-4-isopropylbenzene

≥95.0%

Synonym: 4-Bromocumene

  • CAS Number 586-61-8

  • Empirical Formula (Hill Notation) C9H11Br

  • Molecular Weight 199.09

  •  Beilstein Registry Number 1858096

  •  EC Number 209-577-5

  •  MDL number MFCD00039159

  •  PubChem Substance ID 329748762

Purchase

Properties

Related Categories Aryl, Building Blocks, C9 to C12, Chemical Synthesis, Halogenated Hydrocarbons,
InChI Key   MOZHUOIQYVYEPN-UHFFFAOYSA-N
assay   ≥95.0%
refractive index   n20/D 1.537
bp   103-104 °C(lit.)
density   1.286 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN2810 - class 6.1 - PG 3 - EHS - Toxic, liquids, organic, n.o.s., HI: all
WGK Germany 
2
Flash Point(F) 
201.2 °F
Flash Point(C) 
94 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?