EMAIL THIS PAGE TO A FRIEND
09427 Sigma-Aldrich

1,5-Cyclooctadiene

puriss., ≥99.0% (GC)

  • CAS Number 111-78-4

  • Empirical Formula (Hill Notation) C8H12

  • Molecular Weight 108.18

  •  Beilstein Registry Number 2036542

  •  EC Number 203-907-1

  •  MDL number MFCD00001752

  •  PubChem Substance ID 329748764

Purchase

Properties

Related Categories Alkenes, Building Blocks, Chemical Synthesis, Cyclic, Organic Building Blocks More...
vapor pressure   25.8 mmHg ( 37.7 °C)
  6.8 mmHg ( 25 °C)
InChI Key   VYXHVRARDIDEHS-QGTKBVGQSA-N
grade   puriss.
assay   ≥99.0% (GC)
autoignition temp.   431 °F
refractive index   n20/D 1.493(lit.)
  n20/D 1.494
bp   149-150 °C(lit.)
mp   −69 °C(lit.)
density   0.882 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2520 3 / PGIII
WGK Germany 
3
RTECS 
GX9560000
Flash Point(F) 
100.4 °F
Flash Point(C) 
38 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

246050 1,5-Cyclooctadiene, purified by redistillation, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?