• USA Home
  • 09439 - trans,trans-1,4-Diphenyl-1,3-butadiene

EMAIL THIS PAGE TO A FRIEND
09439 Sigma-Aldrich

trans,trans-1,4-Diphenyl-1,3-butadiene

purum, ≥98.0% (GC)

Synonym: β,β′-Bistyryl, DPB

  • CAS Number 538-81-8

  • Linear Formula C6H5CH=CHCH=CHC6H5

  • Molecular Weight 206.28

  •  Beilstein Registry Number 1905939

  •  MDL number MFCD00004791

  •  PubChem Substance ID 329748765

Purchase

Properties

InChI Key   JFLKFZNIIQFQBS-FNCQTZNRSA-N
grade   purum
assay   ≥98.0% (GC)
bp   350 °C(lit.)
mp   150-152 °C(lit.)
  150-152 °C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D206008 trans,trans-1,4-Diphenyl-1,3-butadiene, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?