EMAIL THIS PAGE TO A FRIEND
09480 Sigma-Aldrich

2-Aminothiazol

technical, ≥90% (NT)

Synonym: 2-Thiazolamine

  • CAS Number 96-50-4

  • Empirical Formula (Hill Notation) C3H4N2S

  • Molecular Weight 100.14

  •  Beilstein Registry Number 105738

  •  EC Number 202-511-6

  •  MDL number MFCD00005325

  •  PubChem Substance ID 329748769

Purchase

Properties

Related Categories Building Blocks, C3 to C7, Chemical Synthesis, Heterocyclic Building Blocks, Thiazoles More...
InChI Key   RAIPHJJURHTUIC-UHFFFAOYSA-N
grade   technical
assay   ≥90% (NT)
impurities   ≤3% water
mp   85-92 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
XJ2100000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?