• USA Home
  • 09495 - 2-Chloro-6-fluorobenzylamine

EMAIL THIS PAGE TO A FRIEND
09495 Sigma-Aldrich

2-Chloro-6-fluorobenzylamine

≥98.0% (GC)

  • CAS Number 15205-15-9

  • Empirical Formula (Hill Notation) C7H7ClFN

  • Molecular Weight 159.59

  •  Beilstein Registry Number 2357724

  •  EC Number 239-259-1

  •  MDL number MFCD00042458

  •  PubChem Substance ID 24846346

Purchase

Properties

Related Categories Amines, Aryl Fluorinated Building Blocks, Asymmetric Synthesis, Building Blocks, C7-C8,
InChI Key   GVULSXIBCHPJEH-UHFFFAOYSA-N
assay   ≥98.0% (GC)
refractive index   n20/D 1.539
bp   91-93 °C/20 mmHg(lit.)
density   1.24 g/mL at 20 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2735PSN1 8 / PGII
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?