• USA Home
  • 09620 - 11-Aminoundecanoic acid

EMAIL THIS PAGE TO A FRIEND
09620 Sigma-Aldrich

11-Aminoundecanoic acid

purum, ≥97.0% (NT)

  • CAS Number 2432-99-7

  • Linear Formula NH2(CH2)10CO2H

  • Molecular Weight 201.31

  •  Beilstein Registry Number 1767291

  •  EC Number 219-417-6

  •  MDL number MFCD00008150

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Linear Core Amino Acids, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   GUOSQNAUYHMCRU-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (NT)
mp   188-191 °C(lit.)
  188-191 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
YQ2293000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

A82605 11-Aminoundecanoic acid, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?