• USA Home
  • 09666 - 1H,1H,2H-Perfluoro-1-decene

EMAIL THIS PAGE TO A FRIEND
09666 Sigma-Aldrich

1H,1H,2H-Perfluoro-1-decene

purum, ≥95.0% (GC)

Synonym: 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluoro-1-decene

  • CAS Number 21652-58-4

  • Linear Formula CF3(CF2)7CH=CH2

  • Molecular Weight 446.10

  •  Beilstein Registry Number 2066558

  •  EC Number 244-503-5

  •  MDL number MFCD00039246

Purchase

Properties

InChI Key   NKAMGQZDVMQEJL-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.301(lit.)
  n20/D 1.301
bp   146-149 °C(lit.)
density   1.677 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
257 °F
Flash Point(C) 
125 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

370576 1H,1H,2H-Perfluoro-1-decene, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?