• USA Home
  • 09683 - Ammonium ion solution for ISE

EMAIL THIS PAGE TO A FRIEND
09683 Sigma-Aldrich

Ammonium ion solution for ISE

0.1 M NH4+, analytical standard (for ion-selective electrodes)

Synonym: Ammonium chloride solution

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ion Standard Solutions for ISE More...
grade   analytical standard (for ion-selective electrodes)
InChI Key   NLXLAEXVIDQMFP-UHFFFAOYSA-N
shelf life   limited shelf life, expiry date on the label
concentration   0.1 M NH4+
density   0.995 g/mL at 25 °C(lit.)

Description

General description

Visit our Sensor Applications portal to learn more.

Preparation Note

prepared with NH4Cl and H2O

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

99978 Ammonium ionophore I - cocktail A, Selectophore
09685 Ammonium ion solution for ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
09877 Ammonium ionophore I, Selectophore, function tested, ≥95.0% (Nonactin, HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?