• USA Home
  • 09755 - Maize powder MZ 0.1 (Bt-11), IRMM certified Reference Material

EMAIL THIS PAGE TO A FRIEND
09755 Sigma-Aldrich

Maize powder MZ 0.1 (Bt-11), IRMM certified Reference Material

BioChemika, for the detection of GMO maize (Bt-11) with PCR

Purchase

Properties

product line   BioChemika
CofA   certificate is enclosed in each package
quality   for the detection of GMO maize (Bt-11) with PCR
composition   genetically modified Bt-11 maize, 0.1%

Price and Availability


Pharma QC
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?