• USA Home
  • 09761 - Calibrator diluent solution

EMAIL THIS PAGE TO A FRIEND
09761 Sigma-Aldrich

Calibrator diluent solution

for the determination of albumin with albumin blue 580 (05497)

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Kits, Fluorescent Probes, Labels, Particles and Stains More...
pH   6.0±0.5
suitability   in accordance for fluorescence
storage temp.   2-8°C

Description

Other Notes

Component to 09753 Albumin Fluorescence Assay Kit

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?