EMAIL THIS PAGE TO A FRIEND
09765 Sigma-Aldrich

Fmoc-D-Lys(Dde)-OH

≥96.0% (HPLC)

Synonym: Nα-Fmoc-Nε-Dde-D-lysine, Nα-Fmoc-Nε-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)ethyl]-D-lysine

  • Empirical Formula (Hill Notation) C31H36N2O6

  • Molecular Weight 532.63

  •  Beilstein Registry Number 9457462

  •  MDL number MFCD01862894

  •  PubChem Substance ID 24846399

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Lysine, Peptide Chemistry More...
InChI Key   ZPSRBXWVBNVFTO-RUZDIDTESA-N
assay   ≥96.0% (HPLC)
storage temp.   −20°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?