EMAIL THIS PAGE TO A FRIEND
09771 Sigma-Aldrich

Fmoc-Thr(PO3BzlH)-OH

≥97.0% (HPLC)

Synonym: Fmoc-O-(benzylphospho)-L-threonine

  • CAS Number 175291-56-2

  • Empirical Formula (Hill Notation) C26H26NO8P

  • Molecular Weight 511.46

  •  Beilstein Registry Number 7557104

  •  MDL number MFCD00797870

  •  PubChem Substance ID 329748819

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Threonine More...
InChI Key   HOFDVXHILSPFNS-OSPHWJPCSA-N
assay   ≥97.0% (HPLC)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?