• USA Home
  • 09808 - Boc-D-allyl-Gly-OH (dicyclohexylammonium) salt

EMAIL THIS PAGE TO A FRIEND
09808 Sigma-Aldrich

Boc-D-allyl-Gly-OH (dicyclohexylammonium) salt

≥98.0% (TLC)

Synonym: (R)-2-(Boc-amino)-4-pentenoic acid (dicyclohexylammonium) salt, Boc-D-allylglycine (dicyclohexylammonium) salt

  • CAS Number 221352-64-3

  • Empirical Formula (Hill Notation) C10H17NO4 · C12H23N

  • Molecular Weight 396.56

  •  MDL number MFCD01321012

  •  PubChem Substance ID 329748839

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Glycine Derivatives, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   VMCGMPITVQIMGK-HMZWWLAASA-N
assay   ≥98.0% (TLC)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?