EMAIL THIS PAGE TO A FRIEND
09827 Sigma-Aldrich

Cyclodecene oxide

≥96.0% (GC)

Synonym: 1,2-Epoxycyclodecane

  • CAS Number 286-94-2

  • Empirical Formula (Hill Notation) C10H18O

  • Molecular Weight 154.25

  •  Beilstein Registry Number 104995

  •  MDL number MFCD01862897

  •  PubChem Substance ID 24846454

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Epoxides, Organic Building Blocks, Oxygen Compounds More...
InChI Key   AFCZSFUPWLSMSZ-UHFFFAOYSA-N
assay   ≥96.0% (GC)
refractive index   n20/D 1.484
density   0.962 g/mL at 20 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?