• USA Home
  • 09837 - (+)-trans-p-Menth-2-ene

EMAIL THIS PAGE TO A FRIEND
09837 Sigma-Aldrich

(+)-trans-p-Menth-2-ene

analytical standard

Synonym: (3S,6R)-3-Isopropyl-6-methylcyclohexene

  • CAS Number 5113-93-9

  • Empirical Formula (Hill Notation) C10H18

  • Molecular Weight 138.25

  •  Beilstein Registry Number 2037958

  •  MDL number MFCD01862899

  •  PubChem Substance ID 329748846

Purchase

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical Standards, Analytical/Chromatography, I-N, Puriss p.a.,
grade   analytical standard
InChI Key   WHNGPXQYYRWQAS-UWVGGRQHSA-N
assay   ≥98.5% (sum of enantiomers, GC)
optical activity   [α]20/D +127.0±2.0°, neat
shelf life   limited shelf life, expiry date on the label
refractive index   n20/D 1.450
bp   55-56 °C/12 mmHg(lit.)
density   0.811 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 2319 3 / PGIII
WGK Germany 
3
Flash Point(F) 
118.4 °F
Flash Point(C) 
48 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?