EMAIL THIS PAGE TO A FRIEND
09847 Sigma-Aldrich

Ammonium bisulfate

BioUltra, ≥99.5% (T)

Synonym: Ammonium hydrogen sulfate, Ammonium sulfate monobasic

Purchase

Properties

Related Categories Molecular Biology, Optimization Reagents, Protein Structural Analysis, Proteomics, X-Ray Crystallography More...
InChI Key   BIGPRXCJEDHCLP-UHFFFAOYSA-N
product line   BioUltra
assay   ≥99.5% (T)
impurities   insoluble matter, passes filter test
solubility   H2O: 1 M at 20 °C clear, colorless
anion traces   chloride (Cl-): ≤5 mg/kg
  phosphate (PO43-): ≤10 mg/kg
cation traces   Al: ≤5 mg/kg
  As: ≤0.1 mg/kg
  Ba: ≤5 mg/kg
  Bi: ≤5 mg/kg
  Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Li: ≤5 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤5 mg/kg
  Zn: ≤5 mg/kg
λ   1 M in H2O
UV absorption   λ: 260 nm Amax: 0.045
  λ: 280 nm Amax: 0.039

Description

Application

Ammonium bisulfate is an ammonium salt of sulfuric acid used in the study of chemical reactions involving ammonium salt sulfur acids.

Price and Availability


Laboratory Gloves

Amplified Detection
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2506 8 / PGII
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?