• USA Home
  • 09868 - 2,6-Dimethyl-6-hepten-2-ol

EMAIL THIS PAGE TO A FRIEND
09868 Sigma-Aldrich

2,6-Dimethyl-6-hepten-2-ol

analytical standard

  • CAS Number 32779-58-1

  • Empirical Formula (Hill Notation) C9H18O

  • Molecular Weight 142.24

  •  Beilstein Registry Number 1742422

  •  MDL number MFCD01862906

  •  PubChem Substance ID 24846476

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
InChI Key   ICHVSYBMEODCOO-UHFFFAOYSA-N
assay   ≥96.0% (GC)
refractive index   n20/D 1.444
density   0.844 g/mL at 20 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?