• USA Home
  • 09882 - Ammonium ionophore I - cocktail B

EMAIL THIS PAGE TO A FRIEND
09882 Sigma-Aldrich

Ammonium ionophore I - cocktail B

Selectophore

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ionophore Cocktails More...
grade   Selectophore
composition   Ammonium ionophore I, 3.50 wt. % (09877)
  Dibutyl sebacate, 32.95 wt. % (84838)
  Poly(vinyl chloride), 0.90 wt. % (81392)
  Potassium tetrakis(4-chlorophenyl)borate, 0.35 wt. % (60591)
  Tetrahydrofuran, 62.30 wt. % (87369)

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Solvent polymeric membrane solution for NH4+-selevtive microelectrodes

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
May form explosive peroxides.
RIDADR 
UN 2056 3 / PGII
WGK Germany 
3
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

09683 Ammonium ion solution for ISE, 0.1 M NH4+, analytical standard (for ion-selective electrodes)
09685 Ammonium ion solution for ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?