• USA Home
  • 09892 - 2,5-Dimethylcyclohexanol

EMAIL THIS PAGE TO A FRIEND
09892 Sigma-Aldrich

2,5-Dimethylcyclohexanol

analytical standard, mixture of isomers

  • CAS Number 3809-32-3

  • Empirical Formula (Hill Notation) C8H16O

  • Molecular Weight 128.21

  •  Beilstein Registry Number 2232616

  •  EC Number 223-281-3

  •  MDL number MFCD00001504

  •  PubChem Substance ID 329748857

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
InChI Key   RUADGOLRFXTMCY-UHFFFAOYSA-N
assay   ≥98.0% (total assay of isomers, GC)
refractive index   n20/D 1.458
density   0.908 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
156.2 °F
Flash Point(C) 
69 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?