EMAIL THIS PAGE TO A FRIEND
09943 Sigma-Aldrich

CDI

purum, ≥95.0% (T)

Synonym: 1,1′-Carbonyldiimidazole

  • CAS Number 530-62-1

  • Empirical Formula (Hill Notation) C7H6N4O

  • Molecular Weight 162.15

  •  Beilstein Registry Number 6826

  •  EC Number 208-488-9

  •  MDL number MFCD00005286

  •  PubChem Substance ID 24846498

Purchase

Properties

Related Categories Carbonyldiimidazole Derivatives, Chemical Synthesis, Coupling, Synthetic Reagents
InChI Key   PFKFTWBEEFSNDU-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (T)
mp   115-122 °C
  117-122 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3263 8 / PGII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?