• USA Home
  • 10067 - α-Amylase from Bacillus licheniformis

EMAIL THIS PAGE TO A FRIEND
10067 Sigma-Aldrich

α-Amylase from Bacillus licheniformis

thermostable amylase, powder, light brown, ≥2 U/mg

Synonym: 1,4-α-D-Glucan-glucanohydrolase

Purchase

Description

Other Notes

Flow system for starch determination; Starch hydrolysis kinetics

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol maltose per minute at pH 6.9 at 25°C (starch acc. to Zulkowsky, Cat. No. 85642, as substrate)

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?