10220 Sigma-Aldrich

Silver nitrate

meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%

DOWNLOAD MSDS (PDF)

Synonym: Nitric acid silver(I) salt

  • CAS Number 7761-88-8

  • Linear Formula AgNO3

  • Molecular Weight 169.87

  •  EC Number 231-853-9

  •  MDL number MFCD00003414

  •  PubChem Substance ID 24846566

Properties

Related Categories Pharmacopoeia, Pharmacopoeia A-Z More...
vapor density   5.8 (vs air)
assay   99.8-100.5%
form   solid
quality   meets analytical specification of Ph. Eur., BP, USP
impurities   Al, Pb, Bi and Cu ions, complies
  acidity or alkalinity, complies
  residual solvents, complies
  ≤0.03% subst. not precipitated by HCl (as SO4)
mp   212 °C (dec.)(lit.)
anion traces   nitrite (NO2-): ≤1000 mg/kg
  sulfate (SO42-): ≤100 mg/kg
cation traces   Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  Pb: ≤20 mg/kg
suitability   complies for appearance of solution

Price and Availability

Customers Also Viewed

Silver nitrate

BioXtra, >99% (titration)

Silver nitrate

meets USP testing specifications

Silver nitrate

99.9999% trace metals basis

Silver nitrate

ACS reagent, ≥99.0%

Silver nitrate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?