Synonym: Butyl methyl ketone
CAS Number 591-78-6
Linear Formula CH3(CH2)3COCH3
Molecular Weight 100.16
Beilstein Registry Number 1737676
EC Number 209-731-1
MDL number MFCD00009482
PubChem Substance ID 24846590
Popular Documents: Specification Sheet (PDF)
| grade | reagent grade |
| vapor pressure | 10 mmHg ( 39 °C) |
| assay | 98% |
| refractive index |
n |
| bp | 127 °C(lit.) |
| mp | −57 °C(lit.) |
| density | 0.812 g/mL at 25 °C(lit.) |
Inhalation is suspected of causing peripheral neuropathy.1
10, 50, 250 g in glass bottle
Customers Also Viewed
200 μg/mL in methanol, analytical standard
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?